Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28, Rabbit anti-Mouse, Clone: D665, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen CD28
Clone D665
Applications Flow Cytometry, Immunohistochemistry, Functional Assay, Blocking Assay
Host Species Rabbit
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014971
View Documents
Novus Biologicals
NBP2810540.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB014972
View Documents
Novus Biologicals
NBP2810540.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

CD28 Monoclonal antibody specifically detects CD28 in Mouse Samples. It is validated for Flow Cytometry, Immunohistochemistry, Functional, Surface Plasmon Resonance, Blocking Assay.
Specifications

Specifications

CD28
Flow Cytometry, Immunohistochemistry, Functional Assay, Blocking Assay
Mouse
Primary
D665
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit