Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28, Rabbit anti-Rat, Clone: JJ316, Novus Biologicals™
Rabbit Monoclonal Antibody
$277.00 - $578.50
Specifications
Antigen | CD28 |
---|---|
Clone | JJ316 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize |
Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay, Blocking Assay |
Description
CD28 Monoclonal specifically detects CD28 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize.Specifications
CD28 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay, Blocking Assay | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Apoptosis, Immunology | |
PBS | |
940 | |
Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
JJ316 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Rat | |
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
CD28 | |
IgG κ | |
Protein A purified | |
Recognises rat CD28 and can trigger T-cell proliferation in vitro and in vivo. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title