Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD284/MD-2 Complex Rat anti-Mouse, BV650, Clone: MTS510, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740615
Description
CD284/MD-2 Complex Monoclonal antibody specifically detects CD284/MD-2 Complex in Mouse samples. It is validated for Flow Cytometry.Specifications
CD284/MD-2 Complex | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
MTS510 | |
Brilliant Violet 650 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title