Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMIGD2 Antibody (953730), DyLight 550, R&D Systems™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB83161R
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMIGD2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TMIGD2 | |
Partial recombinant human CD28H protein (amino acids 23-150) [UniProt Q96BF3] | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
953730 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
Mouse | |
Protein A or G purified | |
RUO | |
126259 | |
Store at 4C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction