Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 Hamster anti-Rat  Mouse , BV421, Clone: Ha2/5, BD Horizon™

Hamster Monoclonal Antibody

Manufacturer:  BD Biosciences 566230

Catalog No. BDB566230


Add to cart

Description

Description

CD29 Monoclonal antibody specifically detects CD29 in samples. It is validated for Flow cytometry.
Specifications

Specifications

CD29
Ha2/5
Brilliant Violet 421
Purified
Hamster
Armenian Hamster IgM, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Itgb1; Integrin β1 chain; Integrin beta-1; VLA-4 subunit beta
Rat glomerular epithelial cells
25μg
Cell Biology
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.