Learn More
ITGB1 Mouse anti-Human, FITC, Clone: MEM-101A, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen CD2901
Catalog No. 50-112-1608
Add to cart
Description
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD29 (beta1 integrin subunit, GPIIa) forms non-covalently linked heterodimers with at least 6 different alpha chains (alpha1-alpha6, CD49a-f) determining the binding properties of beta1 (VLA) integrins. These integrins mediate cell adhesion to collagen, fibronectin, laminin and other extracellular matrix (ECM) components. This interaction hinders cell death, whereas disruption of anchorage to ECM leads to apoptosis. Decreased expression of most beta1 integrins correlates with acquiring multidrug resistance of tumour cells during selection in presence of antitumour drug. In platelets, translocation of intracellular pool of beta1 integrins to the plasma membrane following thrombin stimulation. These integrins are also up-regulated in leukocytes during emigration and extravascular migration and appear to be critically involved in regulating the immune cell trafficking from blood to tissue, as well as in regulating tissue damage and disease symptoms related to inflammatory bowel disease. Through a beta1 integrin-dependent mechanism, fibronectin and type I collagen enhance cytokine secretion of human airway smooth muscle in response to IL-1beta.
Specifications
Specifications
CD29
Monoclonal
FITC
ITGB1
Liquid
ITGB1
Human CD29
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
MEM-101A
PBS with BSA and 0.1% sodium azide
P05556
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB, ITGB1
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
3688