Learn More
CD294 (CRTH2) Rat anti-Human, APC, Clone: BM16, eBioscience™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Manufacturer:  Invitrogen 17294941
Catalog No. 50-112-3115
Add to cart
Description
Description
Description: This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway. Applications Reported: This BM16 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BM16 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered.
The prostaglandin D2 receptor 2 (PTGDR2), also known as CRTH2, is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, PTGDR2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the PTGDR2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that PTGDR2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo.
Specifications
Specifications
CD294 (CRTH2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9Y5Y4
Crth2, Grp45, MGC130436
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BM16
APC
PTGDR2
Liquid
PTGDR2
IgG2a
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
11251
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.