Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 delta Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299673IR

Catalog No. NB062404


Add to cart

Description

Description

CD3 delta Polyclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

CD3 delta
DyLight 755
50mM Sodium Borate
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Produced in rabbits immunized with purified, recombinant CD3 delta (NP_000723.1; Met 1-Ala 105).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary
915
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit