Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD3ε (Early T-Cell Marker) Ab-2, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD3ε (Early T-Cell Marker) Ab-2, Mouse Monoclonal Antibody.

Specifications

Antigen CD3e
Clone PS1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS401R1
View Documents
Thermo Scientific™
MS401R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS401S
View Documents
Thermo Scientific™
MS401S
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS401S0
View Documents
Thermo Scientific™
MS401S0
100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS401S1
View Documents
Thermo Scientific™
MS401S1
500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD3ε (Early T-Cell Marker) Ab-2, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

 

CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in the majority of T cell neoplasms.

Host Species: Mouse

Clone: PS1

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: ε-chain

Immunogen: Recombinant nonglycosylated ε-chain of CD3 protein

Molecular Weight: ∽20 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD3e
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a
PS1
Monoclonal
Murine
T-Cell
Recombinant nonglycosylated epsilon-chain of CD3 protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit