Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human CD3 epsilon, ultra sensitivity (primary amine labeling);HEK293;25UG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CDEH822325UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD3 epsilon, ultra sensitivity (primary amine labeling);25UG;MABSol Biotinylated Human CD3 epsilon (CDE-H8223) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1). It is the biotinylated form of Human CD3 epsilon Protein (Cat # CDE-H5223).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.2 kDa. The protein migrates as 15-16 kDa on a SDS-PAGE gel under reducing (R) condition.;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 g/mL (100 L/well) can bind Biotinylated Human CD3E, His Tag (Cat. No. CDE-H8223) with a linear range of 0.05-2 ng/mL (QC tested).
Catalog No. NC0820086
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.