Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 epsilon, Mouse anti-Human, Clone: C3e/1931, Novus Biologicals™

Mouse Monoclonal Antibody

$156.50 - $345.50

Specifications

Antigen CD3 epsilon
Clone C3e/1931
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010429
View Documents
Novus Biologicals
NBP279706100UG
100 ug
Each for $345.50
Add to cart
 
NB010430
View Documents
Novus Biologicals
NBP27970620UG
20 ug
Each for $156.50
Add to cart
 
Description

Description

CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Human Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

CD3 epsilon
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Human
Primary
C3e/1931
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit