Learn More
CD3 epsilon Mouse anti-Mouse, Alexa Fluor™ 750, Clone: 145-2C11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252693AF750
Catalog No. NP22693A70
Add to cart
Description
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Mouse samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
CD3 epsilon
145-2C11
Western Blot, Flow Cytometry
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Recognises a 25-kDa protein component (CD3e, originally called T3-e) of the antigen-specific T-cell receptor.
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD3E
H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone.
0.1 mL
Primary
916
Mouse