Learn More
CD3 epsilon, Rabbit anti-Human,Mouse,Rat,Porcine,Amphibian,Bovine,Canine,Chicken,Equine,Feline,Guinea Pig,Hamster,Primate,Rabbit,Sheep, Alexa Fluor 532, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP233116AF532
Catalog No. NB005655
Add to cart
Description
Description
CD3 epsilon Polyclonal specifically detects CD3 epsilon in Human, Rat, Primate samples. It is validated for Western Blot.
Specifications
Specifications
CD3 epsilon
Alexa Fluor 532
50mM Sodium Borate with 0.05% Sodium Azide
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Rabbit
IgG
0.1 mL
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary
Recognizes the epsilon-chain of CD3, which consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta) with MW ranging from 16-28kDa.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Western Blot
Purified, Conjugated
CD3E
A synthetic peptide aa156-168 (ERPPPVPNPDYEP) from human CD3e
Protein A purified
RUO
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Equine, Feline, Guinea Pig, Hamster, Primate, Rabbit