Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Antibody (500A2), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252643AF532
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry.Specifications
CD3 epsilon | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD3E | |
T cell receptor complexes derived from mouse C6VL-BS thymoma cells (C57BL/Ka) . | |
Primary | |
The 25-kDa epsilon chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse stains. | |
Store at 4°C in the dark. |
Flow Cytometry | |
500A2 | |
Flow Cytometry | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Rabbit | |
0.1 mL | |
916 | |
Mouse | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction