Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 gamma Mouse anti-Human, Clone: 5B7B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261729

Catalog No. NBP2-61729


Add to cart

Description

Description

CD3 gamma Monoclonal antibody specifically detects CD3 gamma in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

CD3 gamma
5B7B2
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Purified
CD3G
Purified recombinant fragment of human CD3G (AA: extra 23-116) expressed in E. Coli.
20.5 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ79544, FLJ94613, MGC138597, T3G, T-cell antigen receptor complex, gamma subunit of T3, T-cell receptor T3 gamma chain, T-cell surface glycoprotein CD3 gamma chain
Mouse
IgG1
Protein G purified
Apoptosis, Innate Immunity
Primary
917
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit