Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Human , BV421, Clone: OKT3, BD Biosciences™

Mouse Monoclonal Antibody

$182.70 - $387.50

Specifications

Antigen CD3
Clone OKT3
Applications Flow Cytometry
Conjugate Brilliant Violet 421
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566697
View Documents
BD Biosciences
566697
25 Tests
Each for $182.70
Add to cart
 
BDB566696
View Documents
BD Biosciences
566696
100 Tests
Each for $387.50
Add to cart
 
Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD3
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Sheep Erythrocyte Rosette-purified Human T Cells
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimum conditions, and unconjugated antibody and free BD Horizon BV421 were removed.
OKT3
Brilliant Violet 421
Mouse
Human
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE
Mouse BALB/c x A/J, also known as CAF1 IgG2a, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit