Learn More
CD3e Mouse anti-Human, PE, Clone: UCHT1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen GM40135
Catalog No. 50-112-1621
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.
Specifications
Specifications
CD3
Monoclonal
PE
CD3E
Liquid
CD3E
Human CD3e
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
UCHT1
PBS with 1% BSA and 0.05% sodium azide; pH 7.2
P07766
CD3-DELTA; T3D; CD3 antigen; delta subunit; CD3 delta; CD3d antigen; delta polypeptide TiT3 complex; OKT3; delta chain; T-cell receptor T3 delta chain; T-cell surface glycoprotein CD3 delta chain; CD3/TCR complex; HCD3
Mouse
IgG1
1 mL
4° C, store in dark
Primary
916