Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3d Mouse anti-Human, TRI-COLOR®, Clone: 7D6, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MHCD0306

Catalog No. 50-112-1190


Add to cart
This item is not returnable. View return policy

Description

Description

The TRI-COLOR™ conjugate is a tandem R-phycoerythrin (PE)-Cy5™ conjugate that permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.

CD3d is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The CD3d protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in the CD3d gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms of CD3d have been found. Other variants of CD3d may also exist, but the full-length nature of their transcripts have yet to be defined.
Specifications

Specifications

CD3
Monoclonal
TRI-COLOR™
CD3D
Liquid
CD3D
Human CD3 delta
Purified
IVD
Antibody
Monoclonal
Human
Flow Cytometry
7D6
PBS with 10% sucrose, BSA and 0.1% sodium azide
P04234
CD3-DELTA, T3D, CD3 antigen, delta subunit, CD3 delta, CD3d antigen, delta polypeptide TiT3 complex, OKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain, CD3/TCR complex
Mouse
IgG2a
0.5 mL
4° C, store in dark
Primary
915
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit