Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3ε Mouse anti-Porcine, APC, Clone: PPT3, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 451011
Description
SouthernBiotech anti-porcine primary antibodies are available in a variety of conjugated formats including Alexa Fluor.
Specifications
CD3ε | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Purified CD3 molecules from porcine thymus | |
RUO | |
Porcine | |
IgG1 |
Flow Cytometry | |
PPT3 | |
APC | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title