Learn More
CD3 zeta Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: EM-54, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237708AF532
Catalog No. NP237708A32
Add to cart
Description
Description
CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Flow Cytometry.
Specifications
Specifications
CD3 zeta
EM-54
Western Blot, Flow Cytometry, Flow (Intracellular)
CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Antibody recognizes phosphorylated tyrosine 142 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells.
Western Blot, Flow Cytometry
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CD247
CP1 (pTyr142) - KLH.
0.1 mL
Primary
919
Human, Mouse