Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 zeta Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: EM-55, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP237709AF594

Catalog No. NP237709A94


Add to cart

Description

Description

CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Flow Cytometry.
Specifications

Specifications

CD3 zeta
EM-55
Western Blot, Flow Cytometry, Flow (Intracellular)
CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Antibody recognizes phosphorylated tyrosine 111 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells.
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD247
BP1 (pTyr111/123) - KLH.
0.1 mL
Primary
919
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit