Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 zeta Mouse anti-Human, Mouse, Alexa Fluor™ 750, Clone: EM-54, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237708AF750
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).Specifications
CD3 zeta | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CD247 | |
CP1 (pTyr142) - KLH. | |
Primary | |
Antibody recognizes phosphorylated tyrosine 142 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
EM-54 | |
Western Blot, Flow Cytometry, Flow (Intracellular) | |
CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
Mouse | |
0.1 mL | |
919 | |
Human, Mouse | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title