Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Mouse anti-Human, Unconjugated, Clone: CON6D/B5, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CD30
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF19931MI
View Documents
MilliporeSigma
MABF1993100UG
100 μg
Each for $372.00
Add to cart
 
MABF19932MI
View Documents
MilliporeSigma
MABF199325UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

CD30
Unconjugated
Mouse
Tumor necrosis factor receptor superfamily member;CD30L receptor;Ki-1 antigen;Lymphocyte activation antigen CD30
L428 Hodgkin's lymphoma cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin)
Purified
Human
TNFRSF8, CD30
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit