Learn More
CD30/TNFRSF8 Human anti-Human, Alexa Fluor 750, Clone: Hu8, R&D Systems™
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9576S100UG
Catalog No. FAB9576S100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD30/TNFRSF8
Hu8
Flow Cytometry 0.25-1 ug/10^6 cells
Supernatant
Human
IgG1
100 ug
Primary
943
Human
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Human CD30/TNFRSF8
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CD30/TNFRSF8 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Brentuximab. This product is for research use only.