Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30/TNFRSF8 Mouse anti-Human, PE-Cyanine7, Clone: OTI1C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270374PECY7

Catalog No. N270374PEC7


Add to cart

Description

Description

CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD30/TNFRSF8
OTI1C6
Flow Cytometry
Affinity Purified, Conjugated
TNFRSF8
Recombinant protein fragment corresponding to amino acids 19-379 of human CD30/TNFRSF8 (NP_001234) produced in E.coli.
Immunogen affinity purified
Apoptosis, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Embryonic Stem Cell Markers, Immunology, Stem Cell Markers
Primary
943.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit