Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
CD30/TNFRSF8 Rabbit anti-Human, Alexa Fluor™ 350, Clone: Ki-3, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252682AF350
Catalog No. NP22682A30
Description
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
CD30/TNFRSF8
Ki-3
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
The antibody binds to CD30.
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
TNFRSF8
Human CD30.
0.1 mL
Primary
943
Human