Learn More
CD30/TNFRSF8 Rabbit anti-Human, Alexa Fluor™ 532, Clone: Ki-4, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252683AF532
Catalog No. NP22683A32
Add to cart
Description
Description
CD30/TNFRSF8 Monoclonal antibody specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
CD30/TNFRSF8
Ki-4
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
The antibody binds to CD30.
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TNFRSF8
Human CD30.
0.1 mL
Primary
943
Human