Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD30/TNFRSF8 Rabbit anti-Human, Alexa Fluor™ 750, Clone: Ki-4, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252683AF750
Description
CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
CD30/TNFRSF8 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TNFRSF8 | |
Human CD30. | |
Primary | |
The antibody binds to CD30. | |
Store at 4°C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
Ki-4 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
Rabbit | |
0.1 mL | |
943 | |
Human | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title