Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300a (CLM-8) Rat anti-Mouse, Unconjugated, Clone: TX41, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CD300a (CLM-8)
Applications Flow Cytometry, Functional Assay, Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23401MI
View Documents
MilliporeSigma
MABF2340100UL
100 μL
Each for $372.00
Add to cart
 
MABF23402MI
View Documents
MilliporeSigma
MABF234025UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

CD300a (CLM-8) Monoclonal antibody specifically detects CD300a (CLM-8) in Mouse samples. It is validated for Flow Cytometry, Function Assay, Immunofluorescence, Western Blotting.
Specifications

Specifications

CD300a (CLM-8)
Unconjugated
Rat
CMRF35-like molecule 8;CLM-8;CD300 antigen-like family member A;Leukocyte mono-Ig-like receptor 1;Mast cell-derived paired immunoglobulin-like receptor 1;Myeloid-associated immunoglobulin-like receptor 1;MAIR-1;MAIR-I
Ba/F3 cells transfected with murine CLM-8 (MAIR-I/CD300a) fused with the Fc portion of human IgG.
Primary
Stable for 1 year at -20°C from date of receipt.
Flow Cytometry, Functional Assay, Immunofluorescence, Western Blot
Purified
Mouse
Cd300a, Clm8, Lmir1, Mcpir1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit