Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD300a/LMIR1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF2640SP
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Polyclonal specifically detects CD300a/LMIR1 in Human samples. It is validated for Western Blot.Specifications
CD300a/LMIR1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4 | |
Goat | |
Affinity Purified | |
RUO | |
11314 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ELISA | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9UGN4 | |
CD300A | |
Mouse myeloma cell line NS0-derived recombinant human CD300a/LMIR1 Leu18-Gln178 Accession # Q9UGN4 | |
25 μg | |
Primary | |
Detects human CD300a/LMIR1 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) LMIR2, rhLMIR3, rhLMIR5, and rhLMIR6 is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction