Learn More
CD300a/LMIR1 Mouse anti-Human, Alexa Fluor™ 532, Clone: MEM-260, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11058726AF532
Catalog No. N1108726A32
Add to cart
Description
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
CD300a/LMIR1
MEM-260
Western Blot, Flow Cytometry
CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody (clone MEM-260) reacts with CD300a, a 60 kDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells.
Western Blot, Flow Cytometry
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CD300A
HPB acute lymphoid leukemia cell line (NM_007261.2).
0.1 mL
Primary
11314
Human