Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD300b Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568464
Description
Predicted to react with Human samples.
CD300 molecule-like family member b (CD300LB), also known as CLM-7, is a gene located on chromosome 17q25.1, encoding an immunoreceptor that belongs to the CD300 family of molecules. CD300LB is primarily expressed on the surface of myeloid cells, including dendritic cells and macrophages, where it plays an essential role in modulating immune responses. The receptor is involved in the regulation of cell activation and inflammatory responses through its interactions with phosphatidylserine and other lipid ligands, which can lead to either activation or inhibition of immune cell functions. CD300LB influences processes like antigen presentation, phagocytosis, and cytokine production, thereby impacting innate immune responses and the maintenance of immune homeostasis. Its expression and activity are of particular interest in contexts like autoimmunity, infection, and cancer, where it may contribute to disease progression and serve as a potential therapeutic target in modulating immune functions.
Specifications
CD300b | |
Polyclonal | |
Unconjugated | |
CD300LB | |
CD300 antigen like family member B; CD300 antigen-like family member B; CD300 molecule like family member b; CD300 molecule-like family member b; CD300b; CD300LB; CLM7; CLM-7; CMRF35A2; CMRF35-A2; CMRF35-like molecule 7; CMRF-35-like molecule-7; Gm253; Immune receptor expressed on myeloid cells 3; IREM3; IREM-3; Leukocyte mono-Ig-like receptor 5; LMIR5; TREM5; TREM-5; triggering receptor expressed on myeloid cells 5; UNQ2530/PRO6029 | |
Rabbit | |
affinity chromatography | |
RUO | |
124599 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A8K4G0 | |
CD300LB | |
Synthesized peptide derived from human CD300b [aa 61-110]. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction