Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD300c Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553184
Description
Immunogen sequence: PWLRDFHDPI VEVEVSVFPA GTTTASSPQS SMGTSGPPTK LPVHTWPSVT RKDSPEPSPH PGSLFSNVR Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 36%.
The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes (Jackson et al., 1992 [PubMed 1349532]).
Specifications
CD300c | |
Polyclonal | |
Unconjugated | |
CD300C | |
CD300 antigen-like family member C; CD300c; CD300c antigen; CD300c molecule; Clm6; CLM-6; CMRF35; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35A; CMRF-35A; CMRF35A leukocyte immunoglobulin-like receptor; CMRF35A1; CMRF35-A1; CMRF-35-like molecule 6; CMRF35-like molecule 6; Gcom1; IgSF16; immunoglobulin superfamily member 16; LIR; RGD1565161 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08708 | |
CD300C | |
Recombinant protein corresponding to Human CD300c. Recombinant protein control fragment (Product #RP-91029). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction