Learn More
CD300e (IREM-2) Mouse anti-Human, APC, Clone: UP-H2, eBioscience™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 17300742
Catalog No. 50-112-3119
Add to cart
Description
Description
Description: The UP-H2 monoclonal antibody recognizes human IREM-2 (immune receptor expressed in monocytic derived cells) also known as LMIR6, and unofficially CD300E and CD300LE. It is a member of the CMRF-35 family. This 34 kDa Ig-like containing receptor is an activating receptor and through it ft.s charged residues in the cytokplasmic domain interacts with ITAM containing adaptor proteins like DAP12. Expression is found exclusively on monocytes and myeloid DC ft.s. Expression may be transient as differentiation of monocytes leads to down regulation of IREM-2 yet expression is found on mature DCs. It has been also noted that myeloid cell lines such as THP-1 and U937 do not express IREM-2. In vitro addition of UP-H2 antibody either soluble or crosslinked stimulates monocytes. Applications Reported: This UP-H2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UP-H2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered.
CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
Specifications
Specifications
CD300e (IREM-2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q496F6
CMRF35-like molecule 2, CLM2, CD300 antigen-like family member E, CMRF35-A5, Immune receptor expressed on myeloid cells 2, IREM-2, Polymeric immunoglobulin receptor 2, PIgR-2, PIgR2, Poly-Ig receptor 2, CD300LE, CMRF35A5, IREM2, CLM-2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UP-H2
APC
CD300E
Liquid
CD300E
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
342510
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.