Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human CD300e/LMIR6 His Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Supplier: Novus Biologicals™ NBP2518720.5MG
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 18-173 of Human CD300e/LMIR6 The Recombinant Human CD300e/LMIR6 Protein is derived from E. coli. The Recombinant Human CD300e/LMIR6 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH 8.0) | |
TMW: 19.9kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
CD300e/LMIR6 | |
Recombinant |
SDS-PAGE | |
342510 | |
0.5mg | |
CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2 | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction