Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CD300e/LMIR6 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $976.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH 8.0) |
Gene ID (Entrez) | 342510 |
Molecular Weight | TMW: 19.9kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25187201
|
Novus Biologicals™
NBP2518720.1MG |
0.1mg |
Each for $344.00
|
|
NBP25187205
|
Novus Biologicals™
NBP2518720.5MG |
0.5mg |
Each for $976.00
|
|
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 18-173 of Human CD300e/LMIR6 The Recombinant Human CD300e/LMIR6 Protein is derived from E. coli. The Recombinant Human CD300e/LMIR6 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH 8.0) | |
TMW: 19.9kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
CD300e/LMIR6 | |
Yes |
SDS-Page | |
342510 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title