Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300e/LMIR6 Mouse anti-Human, PE/Cy5.5, Clone: UP-H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262230PECY55

Catalog No. N262230PEC5


Add to cart

Description

Description

CD300e/LMIR6 Monoclonal antibody specifically detects CD300e/LMIR6 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD300e/LMIR6
UP-H2
Flow Cytometry
Conjugated, Purified
CD300E
CD300e-HA-transfected cells
>95%, by SDS-PAGE
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody AFP-01 reacts with human alpha-Fetoprotein (AFP), a 70 kDa oncofetal antigen. AFP is a major fetal plasma protein, but is not present in healthy adult tissues. Elevated AFP concentrations in adult plasma may be an early marker of hepatocellular carcinoma or teratoblastoma, while high concentrations in amniotic fluid may indicate severe congenital defects of a fetus.
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2
Mouse
IgG1
0.1 mL
Primary
342510.0
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit