Learn More
CD300f (IREM-1) Mouse anti-Human, eFluor® 660, Clone: UP-D1, eBioscience™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 50300841
 View more versions of this product
Catalog No. 50-112-4331
Add to cart
Description
Description
Description: The monoclonal antibody recognizes human IREM-1 (Immune receptor expressed by myeloid cell 1). IREM-1 is also a member of of the CD300 family specifically referred to as CD300f. IREM-1 is a type I transmembrane protein with a single Ig domain, one predicted N-linked glycosylation site and a long cytoplasmic tail which contains a predicted ITIM domain categorizing it as an inhibitory receptor. Additionally the cytoplasmic domain has been shown to interact with SHP-1 and the p85\alpha subunit of PI3K. Expression is restricted to myeloid cells. In peripheral blood IREM-1 is found on monocytes and granulocytes. In monocyte derived dendritic cells, expression of IREM-1 is downregulated. Applications Reported: This UP-D1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UP-D1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.
Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
Specifications
Specifications
CD300f (IREM-1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8TDQ1
CD300f, CLM-1, CLM1, IREM-1, IREM1, IgSF13, NKIR, CD300 antigen like family member F, CD300 antigen-like family member F, CD300 molecule like family member f, CMRF35-like molecule 1, NK inhibitory receptor, immune receptor expressed on myeloid cells 1, immunoglobin superfamily member 13, immunoglobulin superfam
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UP-D1
eFluor™ 660
CD300LF
Liquid
CD300LF
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
146722
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.