Learn More
CD307c Mouse anti-Human, BV480, Clone: H5, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 746461
Catalog No. BDB746461
Add to cart
Description
Description
CD307c Monoclonal antibody specifically detects CD307c in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD307c
H5
Brilliant Violet 480
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2b, κ
RUO
Primary
Human