Learn More
CD31 Monoclonal Antibody (JC70A), Zeta
Mouse Monoclonal Antibody Cocktail
$215.00 - $340.00
Specifications
Antigen | CD31 |
---|---|
Clone | JC70A |
Concentration | 200 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5914
|
ZETA CORPORATION
Z2136MS |
500 μL |
Each for $215.00
|
|
|||||
50-221-5912
|
ZETA CORPORATION
Z2136ML |
1 mL |
Each for $340.00
|
|
|||||
Description
A recommended positive control tissue for this product is Colon, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD31 | |
200 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Preparation of a spleen from a patient with hairy cell leukemia | |
Primary | |
2-8°C | |
Pecam1 |
JC70A | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P16284 | |
5175 | |
IgG1 κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.