Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31 Mouse anti-Human, PE, Clone: MBC 78.2 (PECAM-1.2; 1.2), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566125
Description
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Flow cytometry.Specifications
CD31 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
MBC 78.2 (also known as PECAM-1.2; 1.2) | |
Vol. Per test: 5μL | |
PECAM-1; PECAM1; EndoCAM; GPIIA' | |
Human CD31 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction