Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 (Endothelial Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.

Supplier:  Epredia RB10333R1

 View more versions of this product

Catalog No. RB10333R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion. Antibody to CD31 is of value in the study of benign and malignant vascular tumors. Staining for CD31 has also been used to measure angiogenesis, which reportedly predicts tumor recurrence.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminus of mouse CD31 protein

Molecular Weight: 100 and 130 kDa

Positive Control: Tonsil, Placenta

Cellular Localization: Endothelial cells in blood vessel of tonsil

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD31 (Endothelial Cell Marker)
Polyclonal
Rabbit
Tonsil
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin)
Unconjugated
Synthetic peptide corresponding to C-terminus of mouse CD31 protein
1 mL
Cancer and Tumor Biology
Human
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit