Learn More
CD31/PECAM-1, Mouse anti-Human,Mouse,Feline,Rabbit, Clone: JC/70A, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP280641
Catalog No. NB013714
Add to cart
Description
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse, Feline, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunocytochemistry, CyTOF-ready, Knockout Validated, Dual RNAscope ISH-IHC.
Specifications
Specifications
CD31/PECAM-1
Monoclonal
1 mg/mL
Western Blot : 1:100-1:500, Flow Cytometry 1:100-1:250, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin : 1:10-1:25, Immunohistochemistry-Frozen, Flow (Cell Surface) : 1:100-1:250, Immunocytochemistry, CyTOF-ready, Knockout Validated, Dual RNAscope ISH-IHC
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Mouse
82.5 kDa
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers
5175
Human, Mouse, Feline, Rabbit
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Immunoassay, In Situ Hybridization (ISH)
JC/70A
Unconjugated
PBS
PECAM1
This CD31/PECAM-1 Antibody (JC/70A) was developed against a membrane preparation of a spleen from a patient with hairy cell leukemia.
Protein G purified
RUO
Primary
Rabbit reactivity is reported in a publication (PMID: 21533193) which cites the use of this monoclonal antibody, clone JC/70A. Mouse reactivity reported in scientific literature (PMID: 29700126).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. After opening store under sterile conditions.
IgG1 κ