Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CD31 Rabbit anti-Human, Unconjugated, Clone: 4E20, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen CD31
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB121625MI
View Documents
MilliporeSigma
ZRB121625UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12164XMI
View Documents
MilliporeSigma
ZRB12164X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

CD31
Unconjugated
Rabbit
Platelet endothelial cell adhesion
GST/His-tagged recombinant fragment corresponding to 85 amino acids from the C-terminal, cytoplasmic domain of human Platelet endothelial cell adhesion molecule (CD31; PECAM-1).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Purified
Human
PECAM1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit