Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Rat anti-Mouse, BUV496, Clone: 390, BD Biosciences™

Rat Monoclonal Antibody

Supplier:  BD Biosciences 741084

Catalog No. BDB741084


Only null left
Add to Cart

Description

Description

CD31 Monoclonal antibody specifically detects CD31 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31
Monoclonal
Brilliant Ultraviolet 496
Platelet endothelial cell adhesion molecule; Pecam1; PECAM-1; Pecam-1
C3H/HeJ mouse hematopoietic progenitor cell line 32D
RUO
Mouse
IgG2a κ
Flow Cytometry
390
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.