Learn More
CD318 (CDCP1) Monoclonal Antibody (CSTEM26), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 12318942
Description
This monoclonal antibody CSTEM26 reacts with human CD318, also known as CUB domain-containing protein 1 (CDCP1) and Trask. Applications Reported: This CSTEM26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM26 antibody has been pre-diluted and tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
CD318 (CDCP1) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9H5V8 | |
Cdcp1 | |
Affinity chromatography | |
RUO | |
64866 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
CSTEM26 | |
PE | |
Cdcp1 | |
9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.