Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD318 (CDCP1) Mouse anti-Human, Clone: CSTEM26, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14318980

 View more versions of this product

Catalog No. 14318980


Add to cart

Description

Description

This monoclonal antibody CSTEM26 reacts with human CD318, also known as CUB domain-containing protein 1 (CDCP1) and Trask. Applications Reported: This CSTEM26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM26 antibody has been tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

CD318 (CDCP1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9H5V8
SIMA135, gp140, CUB domain-containing protein 1, Trask
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CSTEM26
Unconjugated
CDCP1
Liquid
CDCP1
IgG2a, kappa
25 μg
4° C, do not freeze
Primary
64866
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit