Learn More
CD32 Mouse anti-Human, PE, Clone: AT10, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen CD3204
Catalog No. 50-113-7415
Add to cart
Description
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
CD32
Monoclonal
PE
FCGR2A
Liquid
FCGR2A
Human CD32
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
AT10
PBS with BSA, sucrose and 0.1% sodium azide
P12318
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2, Immunoglobulin G Fc receptor II, fc-gamma RII-a, fc-gamma-RIIa, fcRII-a, igG Fc receptor II-a, low affinity immunoglobulin gamma Fc region receptor II-a, Fc gamma Receptor II-a, CD32 receptor 2a
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
2212