Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD321 (F11R) Mouse anti-Human, Clone: WK9, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 14932195

Encompass

 View more versions of this product

Catalog No. 14-932-195


Add to cart

Description

Description

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is also involved in reovirus entry into cells.

CD321 (F11R) Monoclonal antibody specifically detects CD321 (F11R) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Specifications

Specifications

CD321 (F11R)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9Y624
Mouse
10 mg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
WK9
Unconjugated
F11r
9130004G24; AA638916; BV11 antigen; CD321; ESTM33; F11 R; F11 receptor; F11 receptor/F11R; F11r; JAM; Jam1; JAM-1; JAMA; JAM-A; Jcam; Jcam1; junction cell adhesion molecule A; junction cell adhesion molecule1; junctional adhesion molecule 1; Junctional adhesion molecule A; KAT; Ly106; PAM-1; Platelet adhesion molecule 1; platelet F11 receptor; UNQ264/PRO301
Affinity chromatography
RUO
50848
4° C
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit