Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD321 (JAM-1) Rat anti-Mouse, BV510, Clone: H202-106, BD OptiBuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 744912
Description
CD321 (JAM-1) Monoclonal antibody specifically detects CD321 (JAM-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD321 (JAM-1) | |
H202-106 | |
Brilliant Violet 510 | |
Rat | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG1, κ | |
RUO | |
Primary | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title